Face shield, B-line
Leverancier: Bollé Safety
B-Line B-line
BOLEBL20PIEA 11.8 EUR
BOLEBL20PI
Face shield, B-line
Gelaatsschermen
B-LINE is a safety eyewear range available to all and geared to the most common industrial risks.
  • High quality face shields
  • UVA/UVB protection
  • Anti-scratch/anti-fog
Weight: 254 g
Certificaten: EN 166
Leveringsinformatie: Supplied with cord.