Pentafluorobenzoyl chloride acylation reagent, for GC derivatisation

Leverancier: MACHEREY-NAGEL
Gevaar

MANA701120.101EA 333 EUR
MANA701120.101
Pentafluorobenzoyl chloride acylation reagent, for GC derivatisation
Pentafluorobenzoyl chloride

Pentafluorobenzoyl chloride (PFBC) will react with hydroxyls, primary and secondary amines, amides and thiols.


  • Addition of halogenated carbons enhances detectability by GC-ECD
  • Derivatives are hydrolytical stable
  • Increased sensitivity by adding molecular weight

Formule: C₇ClF₅O
MW: 230,52 g/mol
Kookpunt: 158…159 °C (760 mm Hg)
Dichtheid: 1,601 g/cm³ (d20°/4°)
MDL-nummer: MFCD00000657
CAS-nummer: 2251-50-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR