Iron, foil, IRMM®, certified reference material, Thickness: 0.1 mm, Supelco®


M-Clarity™ quality level = MQ100

ECNRM524A-1EAEA 0 EUR
ECNRM524A-1EA
Iron, foil, IRMM®, certified reference material, Thickness: 0.1 mm, Supelco®
Iron
Formule: Fe
MW: 55,85 g/mol
MDL-nummer: MFCD00010999
CAS-nummer: 7439-89-6

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR