Avantor® Hichrom, Titanium Frit
Leverancier: VWR
Avantor® Hichrom
HICHHI-112EA 0 EUR
HICHHI-112
Avantor® Hichrom, Titanium Frit
Filters Voorkolom filters en frits
Titanium frits are resistant to corrosion, biocompatible and recommended for protein analysis.