3-Amino-2-nitropyridine ≥98.0%
Leverancier: TCI EUROPE
|
Waarschuwing
|
TCIAA2149-1GEA
49.5
EUR
TCIAA2149-1G
3-Amino-2-nitropyridine ≥98.0%
3-Amino-2-nitropyridine
Formule:
C₅H₅N₃O₂ MW: 139,11 g/mol |
MDL-nummer:
MFCD00044102 CAS-nummer: 13269-19-7 EINECS: 236-260-9 |
Specificaties testresultaten
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 197.0 to 203.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...