L(+)-allo-Isoleucine ≥95.0%

Leverancier: TCI EUROPE

TCIAA2214-200MGEA 193 EUR
TCIAA2214-200MG
L(+)-allo-Isoleucine ≥95.0%
L(+)-allo-Isoleucine
Formule: H₃CCH₂CH(CH₃)CH(NH₂)COOH
MW: 131,17 g/mol
MDL-nummer: MFCD00066446
CAS-nummer: 1509-34-8
EINECS: 216-142-3

Bestel nu

Specificaties testresultaten

Appearance White to Light yellow powder to crystal
Purity (Nonaqueous Titration) min. 95.0 %
Specific rotation [a]20/D +35.0 ~ +39.0 deg (C=4,6mol/l HCl)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR