2-Amino-3-fluoropyridine ≥98.0% (by GC, titration analysis)

Leverancier: TCI EUROPE
Waarschuwing

TCIAA2799-1GEA 82.5 EUR
TCIAA2799-1G
2-Amino-3-fluoropyridine ≥98.0% (by GC, titration analysis)
2-Amino-3-fluoropyridine
Formule: C₅H₅FN₂
MW: 112,11 g/mol
Kookpunt: 110°C
MDL-nummer: MFCD04114135
CAS-nummer: 21717-95-3

Bestel nu

Specificaties testresultaten

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 42.0 to 46.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR