4-Bromobenzylamine ≥98.0% (by GC, titration analysis)
Leverancier: TCI EUROPE
|
Gevaar
|
TCIAB4713-1GEA
60.5
EUR
TCIAB4713-1G
4-Bromobenzylamine ≥98.0% (by GC, titration analysis)
4-Bromobenzylamine
Formule:
C₇H₈BrN MW: 186,05 g/mol Kookpunt: 125°C Smeltpunt: 26°C Opslagtemperatuur: Koelkast |
MDL-nummer:
MFCD00047931 CAS-nummer: 3959-07-7 EINECS: 223-559-4 UN: 3259 ADR: 8,III |
Specificaties testresultaten
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...