2-Bromo-4-isopropylaniline ≥98.0%

Leverancier: TCI EUROPE
Waarschuwing

TCIAB4724-5GEA 42.9 EUR
TCIAB4724-5G
2-Bromo-4-isopropylaniline ≥98.0%
2-Bromo-4-isopropylaniline
Formule: C₉H₁₂BrN
MW: 214,1 g/mol
Kookpunt: 111°C
MDL-nummer: MFCD00051585
CAS-nummer: 51605-97-1

Bestel nu

Specificaties testresultaten

Appearance Colorless to Yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR