4-(1H-Imidazol-1-yl)benzonitrile ≥98.0%

Leverancier: TCI EUROPE
Gevaar

TCIAC2045-5GEA 179 EUR
TCIAC2045-5G
4-(1H-Imidazol-1-yl)benzonitrile ≥98.0%
4-(1H-Imidazol-1-yl)benzonitrile
Formule: C₁₀H₇N₃
MW: 169,1800 g/mol
MDL-nummer: MFCD00127147
CAS-nummer: 25372-03-6
UN: 3439
ADR: 6.1,III

Bestel nu

Specificaties testresultaten

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 152.0 to 156.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR