1-Cyclohexylpiperidine ≥98.0% (by GC, titration analysis)
Leverancier: TCI EUROPE
|
Waarschuwing
|
TCIAC3032-25MLEA
105
EUR
TCIAC3032-25ML
TCIAC3032-5ML
1-Cyclohexylpiperidine ≥98.0% (by GC, titration analysis)
1-Cyclohexylpiperidine
Formule:
C₁₁H₂₁N MW: 167,29 g/mol Kookpunt: 107°C |
MDL-nummer:
MFCD00014642 CAS-nummer: 3319-01-5 |
Specificaties testresultaten
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...