N-(2',6'-Dimethylphenyl)-2-piperidinecarboxamide ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI EUROPE

TCIAD4348-25GEA 52.8 EUR
TCIAD4348-25G
N-(2',6'-Dimethylphenyl)-2-piperidinecarboxamide ≥98.0% (by HPLC, titration analysis)
N-(2',6'-Dimethylphenyl)-2-piperidinecarboxamide
Formule: C₁₄H₂₀N₂O
MDL-nummer: MFCD01701244
CAS-nummer: 15883-20-2

Bestel nu

Specificaties testresultaten

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 114.0 to 118.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR