Dimethyl pimelate ≥98.0%

Leverancier: TCI EUROPE

TCIAD4476-5GEA 208 EUR
TCIAD4476-5G
Dimethyl pimelate ≥98.0%
Dimethyl pimelate
Formule: C₉H₁₆O₄
MW: 188,22 g/mol
Kookpunt: 122°C
MDL-nummer: MFCD00008470
CAS-nummer: 1732-08-7
EINECS: 217-057-4

Bestel nu

Specificaties testresultaten

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR