4-Iodo-2-nitrotoluene ≥98.0%

Leverancier: TCI EUROPE
Waarschuwing

TCIAI0993-5GEA 81.4 EUR
TCIAI0993-5G
4-Iodo-2-nitrotoluene ≥98.0%
4-Iodo-2-nitrotoluene
Formule: C₇H₆INO₂
MW: 263,03 g/mol
MDL-nummer: MFCD00051090
CAS-nummer: 41252-97-5
EINECS: 255-279-3

Bestel nu

Specificaties testresultaten

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 59.0 to 63.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR