4-Methylpyridazine ≥98.0%

Leverancier: TCI EUROPE
Waarschuwing

TCIAM2343-1GEA 193 EUR
TCIAM2343-1G
4-Methylpyridazine ≥98.0%
4-Methylpyridazine
Formule: C₅H₆N₂
MW: 94,12 g/mol
Kookpunt: 225°C
Vlampunt: 106°C
MDL-nummer: MFCD00011594
CAS-nummer: 1120-88-3

Bestel nu

Specificaties testresultaten

Appearance Colorless to Red to Green clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR