5-Nitro-o-cresol ≥98.0% (by GC, titration analysis)
Leverancier: TCI EUROPE
|
Waarschuwing
|
TCIAN0185-10GEA
59.4
EUR
TCIAN0185-10G
5-Nitro-o-cresol ≥98.0% (by GC, titration analysis)
5-Nitro-o-cresol
Formule:
C₇H₇NO₃ MW: 153,14 g/mol Kookpunt: 180°C |
MDL-nummer:
MFCD00043909 CAS-nummer: 5428-54-6 EINECS: 226-580-7 UN: 2446 ADR: 6.1,III |
Specificaties testresultaten
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 118.0 to 121.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...