Sinapic acid, Supelco®

Leverancier: Merck
Waarschuwing


M-Clarity™ quality level = MQ100

49508-10X10MG-EA 315 EUR
49508-10X10MG-
Sinapic acid, Supelco®
Sinapic acid
Formule: C₁₁H₁₂O₅
MW: 224,21 g/mol
Kookpunt: 378 °C (1013 hPa)
Smeltpunt: 200…202 °C
MDL-nummer: MFCD00004401
CAS-nummer: 530-59-6
EINECS: 208-487-3

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR