N-(Trimethylsilylmethyl)benzylamine ≥96.0% (by GC, titration analysis)
Leverancier: TCI EUROPE
|
Waarschuwing
|
TCIAT3000-5MLEA
35.2
EUR
TCIAT3000-5ML
N-(Trimethylsilylmethyl)benzylamine ≥96.0% (by GC, titration analysis)
N-(Trimethylsilylmethyl)benzylamine
Formule:
C₁₁H₁₉NSi MW: 193,36 g/mol Kookpunt: 71°C Vlampunt: 96°C |
MDL-nummer:
MFCD00012192 CAS-nummer: 53215-95-5 |
Specificaties testresultaten
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 96.0 % |
Purity (Nonaqueous Titration) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...