2',3',4'-Trifluoroacetophenone ≥98.0%
Leverancier: TCI EUROPE
|
Waarschuwing
|
TCIAT3425-1GEA
53.9
EUR
TCIAT3425-1G
2',3',4'-Trifluoroacetophenone ≥98.0%
2',3',4'-Trifluoroacetophenone
Formule:
C₈H₅F₃O MW: 174,12 g/mol |
MDL-nummer:
MFCD00061231 CAS-nummer: 243448-15-9 |
Specificaties testresultaten
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...