Ethyl 4-amino-3-bromo-5-methylbenzoate

Leverancier: Apollo Scientific
Waarschuwing

APOSOR110365-1GEA 14.7 EUR
APOSOR110365-1G
Ethyl 4-amino-3-bromo-5-methylbenzoate
Ethyl 4-amino-3-bromo-5-methylbenzoate
Formule: C₁₀H₁₂BrNO₂
MW: 258,112 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD16631259
CAS-nummer: 1379303-09-9

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR