1-(Bromomethyl)-4-(4-methylphenoxy)benzene

Leverancier: Apollo Scientific

OR110526-1G
APOSOR110526-1GEA 0 EUR
APOSOR110526-1G
1-(Bromomethyl)-4-(4-methylphenoxy)benzene
1-(Bromomethyl)-4-(4-methylphenoxy)benzene
Formule: C₁₄H₁₃BrO
MW: 277,156 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD16619597
CAS-nummer: 78805-82-0
UN: 2811
ADR: 6.1,III

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR