4-(2-Oxopyrrolidin-1-yl)benzenesulphonamide

Leverancier: Apollo Scientific

OR110557-1G OR110557-5G
APOSOR110557-1GEA 222 EUR
APOSOR110557-1G APOSOR110557-5G
4-(2-Oxopyrrolidin-1-yl)benzenesulphonamide
4-(2-Oxopyrrolidin-1-yl)benzenesulphonamide
Formule: C₁₀H₁₂N₂O₃S
MW: 240,279 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00453080
CAS-nummer: 36090-27-4

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR