4-(4-Acetylhomopiperazin-1-yl)aniline

Leverancier: Apollo Scientific

Synoniemen: 4-(4-Acetyl-1,4-diazepan-1-yl)aniline

OR110755-1G
APOSOR110755-1GEA 0 EUR
APOSOR110755-1G
4-(4-Acetylhomopiperazin-1-yl)aniline
4-(4-Acetylhomopiperazin-1-yl)aniline
Formule: C₁₃H₁₉N₃O
MW: 233,309 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD10036744
CAS-nummer: 497249-28-2

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR