N-(5-Methyl-3-phenylisoxazol-4-yl)-N'-(tetrahydrofuran-2-ylmethyl)urea

Leverancier: Apollo Scientific

OR111066-1G
APOSOR111066-1GEA 419 EUR
APOSOR111066-1G
N-(5-Methyl-3-phenylisoxazol-4-yl)-N'-(tetrahydrofuran-2-ylmethyl)urea
N-(5-Methyl-3-phenylisoxazol-4-yl)-N'-(tetrahydrofuran-2-ylmethyl)urea
Formule: C₁₆H₁₉N₃O₃
MW: 301,34 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD28148240
CAS-nummer: 1858255-75-0

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR