4-Nitro-1-(oxolan-2-ylmethyl)-1h-pyrazole

Leverancier: Apollo Scientific

OR111068-1G
APOSOR111068-1GEA 0 EUR
APOSOR111068-1G
4-Nitro-1-(oxolan-2-ylmethyl)-1h-pyrazole
4-Nitro-1-(oxolan-2-ylmethyl)-1h-pyrazole
Formule: C₈H₁₁N₃O₃
MW: 197,191 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD02755679
CAS-nummer: 1006523-68-7

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR