Ethyl 4-bromo-1-benzothiophene-2-carboxylate

Leverancier: Apollo Scientific

Synoniemen: Ethyl 4-bromobenzo[b]thiophene-2-carboxylate

OR111387-1G
APOSOR111387-1GEA 87.3 EUR
APOSOR111387-1G
Ethyl 4-bromo-1-benzothiophene-2-carboxylate
Ethyl 4-bromo-1-benzothiophene-2-carboxylate
Formule: C₁₁H₉BrO₂S
MW: 285,157 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD23699513
CAS-nummer: 93103-82-3

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR