4-Chloro-6-(isopropylamino)-2-(methylsulphanyl)pyrimidine

Leverancier: Apollo Scientific

Synoniemen: 6-Chloro-2-(methylsulphanyl)-N-(prop-2-yl)pyrimidin-4-amine,6-Chloro-N-isopropyl-2-(methylthio)pyrimidin-4-amine

OR11959-1G
APOSOR11959-1GEA 75.7 EUR
APOSOR11959-1G
4-Chloro-6-(isopropylamino)-2-(methylsulphanyl)pyrimidine
4-Chloro-6-(isopropylamino)-2-(methylsulphanyl)pyrimidine
Formule: C₈H₁₂ClN₃S
MW: 217,719 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD09800997
CAS-nummer: 951884-54-1

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR