3-[3-(Chloromethyl)phenyl]-1-methyl-1H-pyrazole, tech

Leverancier: Apollo Scientific
Gevaar

APOSOR12272-250MGEA 0 EUR
APOSOR12272-250MG APOSOR12272-1G
3-[3-(Chloromethyl)phenyl]-1-methyl-1H-pyrazole, tech
3-[3-(Chloromethyl)phenyl]-1-methyl-1H-pyrazole
Formule: C₁₁H₁₁ClN₂
MW: 206,671 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD09702402
CAS-nummer: 912569-61-0
UN: 3265
ADR: 8,III

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR