N-Methyl-3-[(6-methylpyrazin-2-yl)oxy]benzylamine 97%

Leverancier: Apollo Scientific
Gevaar

APOSOR12416-250MGEA 0 EUR
APOSOR12416-250MG APOSOR12416-1G
N-Methyl-3-[(6-methylpyrazin-2-yl)oxy]benzylamine 97%
N-Methyl-3-[(6-methylpyrazin-2-yl)oxy]benzylamine
Formule: C₁₃H₁₅N₃O
MW: 229,278 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD09817528
CAS-nummer: 912569-66-5
UN: 2922
ADR: 8+6.1,III

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR