2-(Thien-2-yl)benzyl alcohol 97%

Leverancier: Apollo Scientific
Waarschuwing

Synoniemen: [2-(Thien-2-yl)phenyl]methanol

OR1433-1G OR1433-250MG
APOSOR1433-1GEA 0 EUR
APOSOR1433-1G APOSOR1433-250MG
2-(Thien-2-yl)benzyl alcohol 97%
2-(Thien-2-yl)benzyl alcohol
Formule: C₁₁H₁₀OS
MW: 190,262 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD06203081
CAS-nummer: 773872-97-2

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR