4-Bromothien-3-yl isocyanate 95%

Leverancier: Apollo Scientific
Gevaar

APOSOR15962-1GEA 498 EUR
APOSOR15962-1G
4-Bromothien-3-yl isocyanate 95%
4-Bromothien-3-yl isocyanate

May form small amount of sediment

Formule: C₅H₂BrNOS
MW: 204,044 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD09879904
CAS-nummer: 937795-88-5
UN: 2206
ADR: 6.1,III

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR