3-Methyl-2-nitrophenylacetic acid

Leverancier: Apollo Scientific
Waarschuwing

Synoniemen: 2-(Carboxymethyl)-6-methylnitrobenzene,3-(Carboxymethyl)-2-nitrotoluene,2-(3-Methyl-2-nitrophenyl)acetic acid

OR18016-1G OR18016-250MG
APOSOR18016-1GEA 0 EUR
APOSOR18016-1G APOSOR18016-250MG
3-Methyl-2-nitrophenylacetic acid
3-Methyl-2-nitrophenylacetic acid
Formule: C₉H₉NO₄
MW: 195,172 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD08276006
CAS-nummer: 18710-86-6

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR