1-(3-Amino-4-nitrophenyl)piperidin-4-ol

Leverancier: Apollo Scientific

OR183521-1G
APOSOR183521-1GEA 147 EUR
APOSOR183521-1G
1-(3-Amino-4-nitrophenyl)piperidin-4-ol
1-(3-Amino-4-nitrophenyl)piperidin-4-ol
Formule: C₁₁H₁₅N₃O₃
MW: 237,255 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD27950443
CAS-nummer: 404009-22-9

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR