5-Amino-4-(3,4-diethoxyphenyl)-1H-pyrazole

Leverancier: Apollo Scientific

Synoniemen: 4-(3,4-Diethoxyphenyl)-2H-pyrazol-3-ylamine

OR1970-1G OR1970-500MG
APOSOR1970-1GEA 0 EUR
APOSOR1970-1G APOSOR1970-500MG
5-Amino-4-(3,4-diethoxyphenyl)-1H-pyrazole
5-Amino-4-(3,4-diethoxyphenyl)-1H-pyrazole
Formule: C₁₃H₁₇N₃O₂
MW: 247,293 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD08056195
CAS-nummer: 1015844-92-4

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR