Benzyl-2-acetamido-2-deoxy-3,6-di-O-benzyl-β-D-glucopyranoside

Leverancier: Apollo Scientific

OR2125T-250MG
APOSOR2125T-250MGEA 0 EUR
APOSOR2125T-250MG
Benzyl-2-acetamido-2-deoxy-3,6-di-O-benzyl-β-D-glucopyranoside
Benzyl-2-acetamido-2-deoxy-3,6-di-O-benzyl-β-D-glucopyranoside
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00210786
CAS-nummer: 119870-30-3

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR