6-Methyl-2,3-dihydropyridazin-3-one hydrate

Leverancier: Apollo Scientific
Waarschuwing

Synoniemen: 6-Methylpyridazin-3(2H)-one hydrate

OR21483-10G OR21483-1G
APOSOR21483-10GEA 0 EUR
APOSOR21483-10G APOSOR21483-1G
6-Methyl-2,3-dihydropyridazin-3-one hydrate
6-Methyl-2,3-dihydropyridazin-3-one hydrate
Formule: C₅H₆N₂O·H₂O
MW: 128,129 g/mol
Smeltpunt: 146…148 °C
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00150586
CAS-nummer: 7143-82-0

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR