2-Bromobenzo[b]furan 97%

Leverancier: Apollo Scientific
Waarschuwing

Synoniemen: 2-Bromo-1-benzofuran,2-Bromobenzofuran

OR23096-10G OR23096-1G
APOSOR23096-10GEA 0 EUR
APOSOR23096-10G APOSOR23096-1G
2-Bromobenzo[b]furan 97%
2-Bromobenzo[b]furan
Formule: C₈H₅BrO
MW: 197,029 g/mol
Kookpunt: 221…223 °C
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD02681987
CAS-nummer: 54008-77-4

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR