4-(Chloromethyl)-2-(2-thienyl)-1,3-thiazole

Leverancier: Apollo Scientific

OR23157-1G OR23157-250MG
APOSOR23157-1GEA 0 EUR
APOSOR23157-1G APOSOR23157-250MG
4-(Chloromethyl)-2-(2-thienyl)-1,3-thiazole
4-(Chloromethyl)-2-(2-thienyl)-1,3-thiazole
Formule: C₈H₆ClNS₂
MW: 215,723 g/mol
Smeltpunt: 51…53 °C
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD01571296
CAS-nummer: 54679-16-2

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR