3-Bromo-N-(5-methylisoxazol-3-yl)propanamide

Leverancier: Apollo Scientific
Waarschuwing

Synoniemen: 3-Bromo-N-(5-methyl-1,2-oxazol-3-yl)propanamide,3-[(3-Bromopropanoyl)amino]-5-methyl-1,2-oxazole

OR23850-500MG
APOSOR23850-500MGEA 0 EUR
APOSOR23850-500MG
3-Bromo-N-(5-methylisoxazol-3-yl)propanamide
3-Bromo-N-(5-methylisoxazol-3-yl)propanamide
Formule: C₇H₉BrN₂O₂
MW: 233,063 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD01763613
CAS-nummer: 349442-99-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR