7-methylchroman-4-one 95+%

Leverancier: Apollo Scientific

Synoniemen: 7-Methylchroman-4-one

OR24523-250MG
APOSOR24523-250MGEA 209 EUR
APOSOR24523-250MG
7-methylchroman-4-one 95+%
7-methylchroman-4-one
Formule: C₁₀H₁₀O₂
MW: 162,185 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00208231
CAS-nummer: 18385-69-8

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR