2-(3-Pyrazin-2-yl-1,2,4-oxadiazol-5-yl)acetohydrazide

Leverancier: Apollo Scientific
Waarschuwing

Synoniemen: 2-(3-Pyrazin-2-yl-1,2,4-oxadiazol-5-yl)ethanohydrazide

OR25658-250MG
APOSOR25658-250MGEA 0 EUR
APOSOR25658-250MG
2-(3-Pyrazin-2-yl-1,2,4-oxadiazol-5-yl)acetohydrazide
2-(3-Pyrazin-2-yl-1,2,4-oxadiazol-5-yl)acetohydrazide
Formule: C₈H₈N₆O₂
MW: 220,188 g/mol
Smeltpunt: 184…186 °C
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD00219748
CAS-nummer: 175203-77-7

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR