cis-4-Phenylpiperidine-3-carboxylic acid, N-BOC protected 99%

Leverancier: Apollo Scientific
Waarschuwing

Synoniemen: cis-1-(tert-Butoxycarbonyl)-4-phenylpiperidine-3-carboxylic acid,cis-1-(tert-Butoxycarbonyl)-3-carboxy-4-phenylpiperidine

OR2630-100MG OR2630-250MG
APOSOR2630-100MGEA 0 EUR
APOSOR2630-100MG APOSOR2630-250MG
cis-4-Phenylpiperidine-3-carboxylic acid, N-BOC protected 99%
cis-4-Phenylpiperidine-3-carboxylic acid, N-BOC protected
Formule: C₁₇H₂₃NO₄
MW: 305,369 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD08457835
CAS-nummer: 197900-84-8

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR