L-Cystine dihydrochloride
Leverancier: Apollo Scientific
APOSBI7950-100GEA
205
EUR
APOSBI7950-100G
L-Cystine dihydrochloride
L-Cystine dihydrochloride
Synthetically derived.
Formule:
C₆H₁₂N₂O₄S₂·2HCl MW: 313,23 g/mol Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00070399 CAS-nummer: 30925-07-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...