(S)-3-(Boc-amino)-3-phenylpropionic acid

Leverancier: Apollo Scientific

APOSBICR121-1GEA 234 EUR
APOSBICR121-1G APOSBICR121-2G APOSBICR121-5G
(S)-3-(Boc-amino)-3-phenylpropionic acid
(S)-3-(Boc-amino)-3-phenylpropionic acid
Formule: C₁₄H₁₉NO₄
MW: 265,31 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD01860892
CAS-nummer: 103365-47-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR