Manumycin A

APOSBIM0135-5MGEA 0 EUR
APOSBIM0135-5MG APOSBIM0135-1MG
Manumycin A
Manumycin A
Formule: C₃₁H₃₈N₂O₇
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00920782
CAS-nummer: 52665-74-4

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR