Polyethyleenglycol 1.000

APOSBIPG1678-250MGEA 0 EUR
APOSBIPG1678-250MG
Polyethyleenglycol 1.000
Polyethyleenglycol 1.000
Formule: H(OCH₂CH₂)nOH
MW: 1000 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00081839
CAS-nummer: 25322-68-3

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR