DL-Tyrosine
Leverancier: Apollo Scientific
|
Waarschuwing
|
APOSBIT0723-1KGEA
481
EUR
APOSBIT0723-1KG
DL-Tyrosine
DL-Tyrosine
Formule:
C₉H₁₁NO₃ MW: 181,19 g/mol Smeltpunt: >300 °C Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00063074 CAS-nummer: 556-03-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...