Rubidium acetate 99%

APOSIN3007-25GEA 0 EUR
APOSIN3007-25G
Rubidium acetate 99%
Rubidium acetate
Formule: H₃CCOORb
MW: 144,51 g/mol
Smeltpunt: 246 °C
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00013058
CAS-nummer: 563-67-7

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR