Rubidium perchlorate 99%

Leverancier: Apollo Scientific
Gevaar

APOSIN3040-25GEA 455 EUR
APOSIN3040-25G
Rubidium perchlorate 99%
Rubidium perchlorate
Formule: ClO₄Rb
MW: 184,92 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00016294
CAS-nummer: 13510-42-4
UN: 1481
ADR: 5.1,II

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR