Terbium(III) bromide hydrate 99.99%

APOSIN3466-50GEA 0 EUR
APOSIN3466-50G APOSIN3466-10G
Terbium(III) bromide hydrate 99.99%
Terbium(III) bromide hydrate
Formule: Br₃Tb·H₂O
MW: 416,65 g/mol
Smeltpunt: 170 °C (ontbinding)
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00049571
CAS-nummer: 15162-98-8

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR